missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetrakis(4-nitrophenyl)-1,4-phenylenediamine 98.0+%, TCI America™

$156.59 - $471.60

Chemical Identifiers

CAS 3283-05-4
Molecular Formula C30H20N6O8
Molecular Weight (g/mol) 592.52
MDL Number MFCD23135517
InChI Key XEUNCVYZWDLKKR-UHFFFAOYSA-N
PubChem CID 76775
IUPAC Name 1-N,1-N,4-N,4-N-tetrakis(4-nitrophenyl)benzene-1,4-diamine
SMILES C1=CC(=CC=C1N(C2=CC=C(C=C2)[N+](=O)[O-])C3=CC=C(C=C3)[N+](=O)[O-])N(C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=C(C=C5)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T34561G
View Documents
TCI America
T34561G
1 g N/A
Only null left
 
T34565G
View Documents
TCI America
T34565G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3283-05-4
592.52
XEUNCVYZWDLKKR-UHFFFAOYSA-N
1-N,1-N,4-N,4-N-tetrakis(4-nitrophenyl)benzene-1,4-diamine
C30H20N6O8
MFCD23135517
76775
C1=CC(=CC=C1N(C2=CC=C(C=C2)[N+](=O)[O-])C3=CC=C(C=C3)[N+](=O)[O-])N(C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=C(C=C5)[N+](=O)[O-]

Specifications

3283-05-4
C30H20N6O8
1 g
C1=CC(=CC=C1N(C2=CC=C(C=C2)[N+](=O)[O-])C3=CC=C(C=C3)[N+](=O)[O-])N(C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=C(C=C5)[N+](=O)[O-]
592.52
592.52
Crystalline Powder
392°C
MFCD23135517
XEUNCVYZWDLKKR-UHFFFAOYSA-N
1-N,1-N,4-N,4-N-tetrakis(4-nitrophenyl)benzene-1,4-diamine
76775
≥98.0% (N)
N,N,N′,N′-Tetrakis(4-nitrophenyl)-1,4-phenylenediamine
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'