missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine 97.0+%, TCI America™

Catalog No. p-7136800
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14871G 1 g
T14875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14871G Supplier TCI America Supplier No. T14871G

Chemical Identifiers

CAS 16858-02-9
Molecular Formula C26H28N6
Molecular Weight (g/mol) 424.55
MDL Number MFCD00036918
InChI Key CVRXLMUYFMERMJ-UHFFFAOYSA-N
Synonym tpen, n,n,n',n'-tetrakis 2-pyridylmethyl ethylenediamine, n1,n1,n2,n2-tetrakis pyridin-2-ylmethyl ethane-1,2-diamine, unii-r9ptu1u29i, n,n,n',n'-tetrakis 2-pyridylmethyl ethane-1,2-diamine, r9ptu1u29i, n,n,n',n'-tetrakis 2-pyridyl-methyl ethylenediamine, 1,2-ethanediamine, n,n,n',n'-tetrakis 2-pyridinylmethyl-, oc-6-11, 1,2-ethanediamine,n,n,n',n'-tetrakis 2-pyridinylmethyl, 1,2-ethanediamine, n,n,n',n'-tetrakis 2-pyridinylmethyl
PubChem CID 5519
IUPAC Name (2-{bis[(pyridin-2-yl)methyl]amino}ethyl)bis[(pyridin-2-yl)methyl]amine
SMILES C(CN(CC1=CC=CC=N1)CC1=CC=CC=N1)N(CC1=CC=CC=N1)CC1=CC=CC=N1

Specifications

Melting Point 112°C
Quantity 1 g
Formula Weight 424.55
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine