missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine 98.0+%, TCI America™

Catalog No. T078125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T078125G 25 g
T0781500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T078125G Supplier TCI America Supplier No. T078125G

Chemical Identifiers

CAS 102-60-3
Molecular Formula C14H32N2O4
Molecular Weight (g/mol) 292.42
MDL Number MFCD00004534
InChI Key NSOXQYCFHDMMGV-UHFFFAOYSA-N
Synonym quadrol, edetol, n,n,n',n'-tetrakis 2-hydroxypropyl ethylenediamine, entprol, adeka quadrol, quadrol l, neutrol te, 1,1',1,1'-ethylenedinitrilo tetra-2-propanol, edetolum latin, 2-propanol, 1,1',1,1'-1,2-ethanediyldinitrilo tetrakis
PubChem CID 7615
IUPAC Name 1-[2-[bis(2-hydroxypropyl)amino]ethyl-(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(CN(CCN(CC(C)O)CC(C)O)CC(C)O)O

Specifications

Color Colorless
Quantity 25 g
Formula Weight 292.42
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetrakis(2-hydroxypropyl)ethylenediamine
EINECSNumber (2)-2410
RTECSNumber UB5604000
TSCA Yes