missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetrakis(2-hydroxyethyl)adipamide 98.0+%, TCI America™
$76.43 - $231.14
Chemical Identifiers
| CAS | 6334-25-4 |
|---|---|
| Molecular Formula | C14H28N2O6 |
| Molecular Weight (g/mol) | 320.39 |
| MDL Number | MFCD07787618 |
| InChI Key | OKRNLSUTBJUVKA-UHFFFAOYSA-N |
| PubChem CID | 95283 |
| IUPAC Name | N,N,N',N'-tetrakis(2-hydroxyethyl)hexanediamide |
| SMILES | OCCN(CCO)C(=O)CCCCC(=O)N(CCO)CCO |
Chemical Identifiers
| 6334-25-4 | |
| 320.39 | |
| OKRNLSUTBJUVKA-UHFFFAOYSA-N | |
| N,N,N',N'-tetrakis(2-hydroxyethyl)hexanediamide |
| C14H28N2O6 | |
| MFCD07787618 | |
| 95283 | |
| OCCN(CCO)C(=O)CCCCC(=O)N(CCO)CCO |
Specifications
| 6334-25-4 | |
| White-Yellow | |
| MFCD07787618 | |
| N,N,N′C,N′C-Tetrakis(2-hydroxyethyl)hexanediamide | |
| OCCN(CCO)C(=O)CCCCC(=O)N(CCO)CCO | |
| 320.39 | |
| 320.39 | |
| Crystalline Powder |
| 128°C | |
| C14H28N2O6 | |
| 5 g | |
| OKRNLSUTBJUVKA-UHFFFAOYSA-N | |
| N,N,N',N'-tetrakis(2-hydroxyethyl)hexanediamide | |
| 95283 | |
| ≥98.0% (GC) | |
| N,N,N′,N′-Tetrakis(2-hydroxyethyl)adipamide |
Safety and Handling
EINECSNumber : (2)-3798
TSCA : No