missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetraethyl-1,3-propanediamine, 97%

Catalog No. AAA1768006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1768006 5 g
AAA1768014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1768006 Supplier Thermo Scientific Chemicals Supplier No. A1768006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 60558-96-5 | C11H26N2 | 186.343 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60558-96-5
Molecular Formula C11H26N2
Molecular Weight (g/mol) 186.343
MDL Number MFCD00009057
InChI Key RCZLVPFECJNLMZ-UHFFFAOYSA-N
Synonym n,n,n',n'-tetraethyl-1,3-propanediamine, n,n,n',n'-tetraethyl trimethylene diamine, 1,3-propanediamine, n,n,n',n'-tetraethyl, acmc-20ak9w, 1,3-bis diethylamino propane, 3-diethylamino propyl diethylamine, n1,n1,n3,n3-tetraethylpropane-1,3-diamine, n,n,n,n-tetraethyl-1,3-propanediamine, the 1,3-propanediamine,n1,n1,n3,n3-tetraethyl-,
PubChem CID 143710
IUPAC Name N,N,N',N'-tetraethylpropane-1,3-diamine
SMILES CCN(CC)CCCN(CC)CC

Specifications

Density 0.81
Boiling Point 133°C (80 mmHg)
Flash Point 76°C (168°F)
Refractive Index 1.4377
Quantity 5 g
UN Number UN2735
Beilstein 1741257
Formula Weight 186.34
Percent Purity 97%
Chemical Name or Material N,N,N',N'-Tetraethyl-1,3-propanediamine
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only