missing translation for 'onlineSavingsMsg'
Learn More

N,N',N″,N‴-Tetraacetylglycoluril 98.0+%, TCI America™

Catalog No. p-7136535
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09855G 5 g
T098525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09855G Supplier TCI America Supplier No. T09855G
Only null left

Chemical Identifiers

CAS 10543-60-9
Molecular Formula C12H14N4O6
Molecular Weight (g/mol) 310.27
MDL Number MFCD00022618
InChI Key KIHGYZTVBURVBA-UHFFFAOYSA-N
Synonym n,n',n,n'-tetraacetylglycoluril, 1,3,4,6-tetraacetyltetrahydroimidazo 4,5-d imidazole-2,5 1h,3h-dione, imidazo 4,5-d imidazole-2,5 1h,3h-dione, 1,3,4,6-tetraacetyltetrahydro, n,n',n,n'-tetraacetylacetyleneurea, n,n',n,n'-tetraacetylglycouril, 1,3,4,6-tetraacetyl-dihydro-1,3 diazolo 4,5-d imidazole-2,5-dione, tetraacetylglycoluril, tetraacetylglykoluril
PubChem CID 82717
IUPAC Name 1,3,4,6-tetraacetyl-octahydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione
SMILES CC(=O)N1C2C(N(C(C)=O)C1=O)N(C(C)=O)C(=O)N2C(C)=O

Specifications

Melting Point 237°C
Color White
Quantity 5 g
Formula Weight 310.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′,N′′,N′′′-Tetraacetylglycoluril
EINECSNumber (5)-1115
TSCA Yes