missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N″,N″-Pentamethyldiethylenetriamine 99.0+%, TCI America™

Catalog No. p-7134824
Change view
Click to view available options
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
P088125ML 25 mL
P0881100ML 100 mL
P0881500ML 500 mL
3 options

Catalog No. P088125ML

Supplier: TCI America P088125ML

Only null left

Chemical Identifiers

CAS 3030-47-5
Molecular Formula C9H23N3
Molecular Weight (g/mol) 173.304
MDL Number MFCD00014876
InChI Key UKODFQOELJFMII-UHFFFAOYSA-N
Synonym pentamethyldiethylenetriamine, 1,1,4,7,7-pentamethyldiethylenetriamine, pmdt, n,n,n',n,n-pentamethyldiethylenetriamine, pmdeta, pmdta, pentamethyldiethylenetriaminek, bis 2-dimethylaminoethyl methyl amine, 2,5,8-trimethyl-2,5,8-triazanonane, n,n',n-pentamethyldiethylenetriamine
PubChem CID 18196
ChEBI CHEBI:39475
IUPAC Name N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine
SMILES CN(C)CCN(C)CCN(C)C

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 86°C
Quantity 25 mL
UN Number 2922
Formula Weight 173.30
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine
EINECSNumber (2)-0147
RTECSNumber IE2100000
TSCA Yes