missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N″,N″-Pentamethyldiethylenetriamine 99.0+%, TCI America™

Catalog No. P088125ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P088125ML 25 mL
P0881100ML 100 mL
P0881500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P088125ML Supplier TCI America Supplier No. P088125ML

Chemical Identifiers

CAS 3030-47-5
Molecular Formula C9H23N3
Molecular Weight (g/mol) 173.304
MDL Number MFCD00014876
InChI Key UKODFQOELJFMII-UHFFFAOYSA-N
Synonym pentamethyldiethylenetriamine, 1,1,4,7,7-pentamethyldiethylenetriamine, pmdt, n,n,n',n,n-pentamethyldiethylenetriamine, pmdeta, pmdta, pentamethyldiethylenetriaminek, bis 2-dimethylaminoethyl methyl amine, 2,5,8-trimethyl-2,5,8-triazanonane, n,n',n-pentamethyldiethylenetriamine
PubChem CID 18196
ChEBI CHEBI:39475
IUPAC Name N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine
SMILES CN(C)CCN(C)CCN(C)C

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 86°C
Quantity 25 mL
UN Number 2922
Formula Weight 173.30
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine
EINECSNumber (2)-0147
RTECSNumber IE2100000
TSCA Yes