missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Ethylenediaminetetrakis(methylenephosphonic Acid) 98.0+%, TCI America™
$93.21 - $564.81
Chemical Identifiers
| CAS | 1429-50-1 |
|---|---|
| Molecular Formula | C6H20N2O12P4 |
| Molecular Weight (g/mol) | 436.123 |
| MDL Number | MFCD00058893 |
| InChI Key | NFDRPXJGHKJRLJ-UHFFFAOYSA-N |
| Synonym | edtmp, edtpo, ethylenebis nitrilodimethylene tetraphosphonic acid, unii-v4cp8rsx7v, v4cp8rsx7v, ethylenediamine tetrakis methylene phosphonic acid, ethane-1,2-diylbis azanetriyl tetrakis methylene tetraphosphonic acid, ethylenedinitrilo-tetramethylenephosphonic acid, ethylenediamine tetramethylenephosphonic acid, n,n,n',n'-ethylenediaminetetrakis methylenephosphonic acid |
| PubChem CID | 15025 |
| IUPAC Name | [2-[bis(phosphonomethyl)amino]ethyl-(phosphonomethyl)amino]methylphosphonic acid |
| SMILES | C(CN(CP(=O)(O)O)CP(=O)(O)O)N(CP(=O)(O)O)CP(=O)(O)O |
Chemical Identifiers
| 1429-50-1 | |
| 436.123 | |
| NFDRPXJGHKJRLJ-UHFFFAOYSA-N | |
| 15025 | |
| C(CN(CP(=O)(O)O)CP(=O)(O)O)N(CP(=O)(O)O)CP(=O)(O)O |
| C6H20N2O12P4 | |
| MFCD00058893 | |
| edtmp, edtpo, ethylenebis nitrilodimethylene tetraphosphonic acid, unii-v4cp8rsx7v, v4cp8rsx7v, ethylenediamine tetrakis methylene phosphonic acid, ethane-1,2-diylbis azanetriyl tetrakis methylene tetraphosphonic acid, ethylenedinitrilo-tetramethylenephosphonic acid, ethylenediamine tetramethylenephosphonic acid, n,n,n',n'-ethylenediaminetetrakis methylenephosphonic acid | |
| [2-[bis(phosphonomethyl)amino]ethyl-(phosphonomethyl)amino]methylphosphonic acid |
Specifications
| 1429-50-1 | |
| <2 (1% H2O soln. 25°C) | |
| MFCD00058893 | |
| edtmp, edtpo, ethylenebis nitrilodimethylene tetraphosphonic acid, unii-v4cp8rsx7v, v4cp8rsx7v, ethylenediamine tetrakis methylene phosphonic acid, ethane-1,2-diylbis azanetriyl tetrakis methylene tetraphosphonic acid, ethylenedinitrilo-tetramethylenephosphonic acid, ethylenediamine tetramethylenephosphonic acid, n,n,n',n'-ethylenediaminetetrakis methylenephosphonic acid | |
| C(CN(CP(=O)(O)O)CP(=O)(O)O)N(CP(=O)(O)O)CP(=O)(O)O | |
| 436.123 | |
| 436.12 | |
| Crystalline Powder |
| White | |
| C6H20N2O12P4 | |
| 25 g | |
| NFDRPXJGHKJRLJ-UHFFFAOYSA-N | |
| [2-[bis(phosphonomethyl)amino]ethyl-(phosphonomethyl)amino]methylphosphonic acid | |
| 15025 | |
| ≥98.0% (T) | |
| N,N,N′,N′-Ethylenediaminetetrakis(methylenephosphonic Acid) |
Safety and Handling
EINECSNumber : (2)-1873
TSCA : Yes