missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Ethylenediaminetetrakis(methylenephosphonic Acid) 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| pH | <2 (1% H2O soln. 25°C) |
| Quantity | 25 g |
| Formula Weight | 436.12 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N,N,N′,N′-Ethylenediaminetetrakis(methylenephosphonic Acid) |
Safety and Handling
| EINECSNumber | (2)-1873 |
| TSCA | Yes |