missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Methylenebisacrylamide 98.0+%, TCI America™

Catalog No. p-7133487
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M287725G 25 g
M2877500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M287725G Supplier TCI America Supplier No. M287725G
Only null left

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
MDL Number MFCD00008625
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C

Specifications

Melting Point 300°C
Color White
Quantity 25 g
Formula Weight 154.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Methylenebisacrylamide
EINECSNumber (2)-1020
RTECSNumber AS3678000
TSCA Yes