missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Methylenebisacrylamide 98.0+%, TCI America™

Supplier:  TCI America M0506100G

 View more versions of this product

Catalog No. M0506100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N,N′-Methylenebisacrylamide [for Electrophoresis]
300°C
C7H10N2O2
100 g
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
N-[(prop-2-enoylamino)methyl]prop-2-enamide
8041
≥98.0% (T)
110-26-9
White
MFCD00008625
n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide
C=CC(=O)NCNC(=O)C=C
154.169
154.17
Crystalline Powder

Chemical Identifiers

110-26-9
154.169
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
8041
C=CC(=O)NCNC(=O)C=C
C7H10N2O2
MFCD00008625
n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide
N-[(prop-2-enoylamino)methyl]prop-2-enamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1020

RTECSNumber : AS3678000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions