missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis

CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol

$99.26 - $239.66

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
MDL Number MFCD00008625
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym MBA
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC164790250
SDS View Documents
Thermo Scientific Chemicals
164790250
25 g Plastic Bottle
Each for $99.26
Only null left
 
AC164791000
SDS View Documents
Thermo Scientific Chemicals
164791000
100 g Plastic Bottle
Each for $239.66
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

110-26-9
154.169
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
8041
C=CC(=O)NCNC(=O)C=C
C7H10N2O2
MFCD00008625
MBA
N-[(prop-2-enoylamino)methyl]prop-2-enamide

Specifications

(1% aq. soln.) (1 cm cell), (1% aq. soln.) (1 cm cell) at 0nm, 0.2 max. at 290nm
0.02% max.
99.0
5.31 (Air = 1.0)
5 (2.5% min. aq. soln.)
Authentic
99+%
(H2C=CHCONH)2CH2
MBA
C=CC(=O)NCNC(=O)C=C
154.169
154.17
Biochemical
Fine Needle-like Crystals or Crystalline Powder
110-26-9
100.0
White
25 g
Plastic Bottle
C7H10N2O2
MFCD00008625
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
N-[(prop-2-enoylamino)methyl]prop-2-enamide
8041
≥99.0%
N, N';-Methylenebisacrylamide, For electrophoresis, For biochemistry
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

RUO – Research Use Only