missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Absorbance | (1% aq. soln.) (1 cm cell), (1% aq. soln.) (1 cm cell) at 0nm, 0.2 max. at 290nm |
| Physical Form | Fine Needle-like Crystals or Crystalline Powder |
| Acrylic Acid | 0.02% max. |
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Density | 5.31 (Air = 1.0) |
| Color | White |
| pH | 5 (2.5% min. aq. soln.) |
| Quantity | 100 g |
| Infrared Spectrum | Authentic |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement: Causes serious eye irritation. Causes skin irritation. Harmful if inhaled. Harmful in contact with skin. May cause respiratory irritation. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not eat, drink or smoke when using this product. IF exposed or concerned: Get medical advice/attention. IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only