missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis

Catalog No. AC164790250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164790250 25 g Plastic Bottle
AC164791000 100 g Plastic Bottle
AC164795000 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164790250 Supplier Thermo Scientific Chemicals Supplier No. 164790250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
MDL Number MFCD00008625
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym MBA
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C

Specifications

Absorbance (1% aq. soln.) (1 cm cell), (1% aq. soln.) (1 cm cell) at 0nm, 0.2 max. at 290nm
Physical Form Fine Needle-like Crystals or Crystalline Powder
Acrylic Acid 0.02% max.
CAS Min % 99.0
CAS Max % 100.0
Density 5.31 (Air = 1.0)
Color White
pH 5 (2.5% min. aq. soln.)
Quantity 25 g
Infrared Spectrum Authentic
Packaging Plastic Bottle
Assay Percent Range 99+%
Linear Formula (H2C=CHCONH)2CH2
Formula Weight 154.17
Percent Purity ≥99.0%
Grade Biochemical
Chemical Name or Material N, N';-Methylenebisacrylamide, For electrophoresis, For biochemistry
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only