missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N'-Methylenebisacrylamide, 99+%

Catalog No. AA4370114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4370114 25 g
AA4370122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4370114 Supplier Thermo Scientific Chemicals Supplier No. 04370114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol

Bisacrylamide is used in biochemistry as a preparatory agent for polyacrylamide gel for electrophoretic separation of nucleic acids and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
MDL Number MFCD00008625
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C

Specifications

Melting Point >300°C
Color White
Density 1.235 g/mL
Quantity 25 g
Beilstein 1706297
Solubility Information It is soluble in water (20g/L) at 20°C,ethanol,and methanol.
Formula Weight 154.17
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material N,N'-Methylenebisacrylamide
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 203-750-9
RTECSNumber AS3678000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only