missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Methylenebisacrylamide, 97%

Catalog No. AAL0469422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0469422 100 g
AAL0469436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0469422 Supplier Thermo Scientific Chemicals Supplier No. L0469422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol

N,N'-Methylenebisacrylamide is used as an intermediate and cross-linking agent utilized in the preparation of polymers like polyacrylamide. It is also used in biochemistry for the preparation of polyacrylamide gels, which is useful for the electrophoretic separation of nucleic acids and proteins. Further, it is employed in the synthesis of acrylic resin and adhesives. In addition to this, it is used as a plugging agent in oilfield drilling and construction grouting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Methylenebisacrylamide is used as an intermediate and cross-linking agent utilized in the preparation of polymers like polyacrylamide. It is also used in biochemistry for the preparation of polyacrylamide gels, which is useful for the electrophoretic separation of nucleic acids and proteins. Further, it is employed in the synthesis of acrylic resin and adhesives. In addition to this, it is used as a plugging agent in oilfield drilling and construction grouting.

Solubility
Soluble in water, ethanol and methanol.

Notes
Air and light sensitive. Store in a cool place. Incompatible with acids, bases, oxidizing agents, reducing agents, iron, iron salts, copper, aluminum, brass and free radical initiators.

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
MDL Number MFCD00008625
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C

Specifications

Melting Point >300°C
Density 1.235
Quantity 100 g
Beilstein 1706297
Solubility Information Soluble in water,ethanol and methanol.
Formula Weight 154.17
Percent Purity 97%
Chemical Name or Material N,N'-Methylenebisacrylamide
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 203-750-9
RTECSNumber AS3678000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only