missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N'-Methylenebisacrylamide, 2% soln.

CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol

Supplier:  Thermo Scientific Chemicals J63265AP

 View more versions of this product

Catalog No. AAJ63265AP


Only null left
Explore available promotions

Description

Description

Bis-acrylamide cross-links with acrylamide to form polyacrylamide gels for electrophoresis. Changing the ratio of bis-acrylamide to acrylamide controls the pore size of polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Bis-acrylamide cross-links with acrylamide to form polyacrylamide gels for electrophoresis
  • Changing the ratio of bis-acrylamide to acrylamide controls the pore size of polyacrylamide gels
Specifications

Specifications

N,N'-Methylenebisacrylamide
Colorless
MFCD00008625
n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide
C=CC(=O)NCNC(=O)C=C
154.169
154.17
Liquid
110-26-9
C7H10N2O2
500 mL
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
N-[(prop-2-enoylamino)methyl]prop-2-enamide
8041
2% soln.

Chemical Identifiers

110-26-9
154.169
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
8041
C=CC(=O)NCNC(=O)C=C
C7H10N2O2
MFCD00008625
n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide
N-[(prop-2-enoylamino)methyl]prop-2-enamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-750-9

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only