missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Methylene-Bis-Acrylamide, Ultra Pure, 5g, MP Biomedicals

110-26-9 Purity: 99.9% (290nm): <<0.2 Acrylic acid content: <<0.02% High purity cross-linker used for precise, critical PAGE gels. May be used in UV scanning gels

Supplier:  MP Biomedicals Inc 04800172

Catalog No. ICN800172


Only null left

Description

Description

Specifications

Specifications

N, N«-Methylene-Bis-Acrylamide
5 g
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
N-[(prop-2-enoylamino)methyl]prop-2-enamide
8041
110-26-9
C7H10N2O2
C=CC(=O)NCNC(=O)C=C
154.169

Chemical Identifiers

110-26-9
154.169
8041
C=CC(=O)NCNC(=O)C=C
C7H10N2O2
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
N-[(prop-2-enoylamino)methyl]prop-2-enamide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions