missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Methylene-bis-Acrylamide, Molecular Biology Grade, Hoefer™

Catalog No. p-3605758
Encompass
Change view
Click to view available options
Quantity:
100 g
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HBGR142500 500 mL
03500525 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. HBGR142500 Supplier Hoefer Inc Supplier No. GR142 500
Cross-linking agent used in forming polyacrylamide gels.

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym bis-Acrylamide
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C

Specifications

Physical Form Crystalline Powder
Color White
Quantity 500 mL
Assay Percent Range ≥99.9%
Linear Formula (CH2CHCONH2)CH2
Formula Weight 154.17
Grade Molecular Biology
Chemical Name or Material N, N'-Methylene-bis-Acrylamide