missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Ditridecyl-3,4,9,10-perylenetetracarboxylic Diimide 98.0+%, TCI America™

Catalog No. p-7128012
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3947200MG 200 mg
D39471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3947200MG Supplier TCI America Supplier No. D3947200MG

Chemical Identifiers

CAS 95689-92-2
Molecular Formula C50H62N2O4
Molecular Weight (g/mol) 755.056
MDL Number MFCD00134587
InChI Key XCAZCFDCJHGAIT-UHFFFAOYSA-N
Synonym PTCDI-C13
PubChem CID 3295215
SMILES CCCCCCCCCCCCCN1C(=O)C2=C3C(=CC=C4C3=C(C=C2)C5=C6C4=CC=C7C6=C(C=C5)C(=O)N(C7=O)CCCCCCCCCCCCC)C1=O

Specifications

Quantity 200 mg
Formula Weight 755.06
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Ditridecyl-3,4,9,10-perylenetetracarboxylic Diimide