missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dipropylethylamine 98.0+%, TCI America™

Catalog No. e083925ml
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E083925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E083925ML Supplier TCI America Supplier No. E083925ML

Chemical Identifiers

CAS 20634-92-8
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.25
MDL Number MFCD00048685
InChI Key XWCCTMBMQUCLSI-UHFFFAOYSA-N
Synonym N-Ethyldipropylamine
PubChem CID 519799
IUPAC Name ethyldipropylamine
SMILES CCCN(CC)CCC

Specifications

Color Colorless
Boiling Point 132°C
Quantity 25 mL
UN Number 2733
Formula Weight 129.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dipropylethylamine