missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenylethylenediamine 98.0+%, TCI America™

Catalog No. D088725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D088725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D088725G Supplier TCI America Supplier No. D088725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150-61-8
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.30
MDL Number MFCD00003019
InChI Key NOUUUQMKVOUUNR-UHFFFAOYSA-N
Synonym 1,2-dianilinoethane, n,n'-diphenylethylenediamine, stabilite, n,n'-ethylenedianiline, nodx, n1,n2-diphenylethane-1,2-diamine, sym-diphenylethylenediamine, 1,2-ethanediamine, n,n'-diphenyl, aniline, n,n'-ethylenedi, n,n'-difenylethylendiamin
PubChem CID 67422
IUPAC Name N1,N2-diphenylethane-1,2-diamine
SMILES C(CNC1=CC=CC=C1)NC1=CC=CC=C1

Specifications

Melting Point 66°C
Boiling Point 228°C
Quantity 25 g
Formula Weight 212.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diphenylethylenediamine
EINECSNumber (9)-2048
RTECSNumber KV4800000
TSCA Yes