missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenyl-N,N'-di(m-tolyl)-1,4-phenylenediamine 98.0+%, TCI America™

Catalog No. D49305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49305G 5 g
D49301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49305G Supplier TCI America Supplier No. D49305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80223-29-6
Molecular Formula C32H28N2
Molecular Weight (g/mol) 440.59
InChI Key ZWZXDJMNYGRYNP-UHFFFAOYSA-N
PubChem CID 14250518
IUPAC Name 1-N,4-N-bis(3-methylphenyl)-1-N,4-N-diphenylbenzene-1,4-diamine
SMILES CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC(=C5)C

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 5 g
Formula Weight 440.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diphenyl-N,N′-di(m-tolyl)-1,4-phenylenediamine