missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenyl-N,N'-bis[4'-(diphenylamino)biphenyl-4-yl]benzidine 98.0+%, TCI America™

Catalog No. D48631G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D48631G Supplier TCI America Supplier No. D48631G
Only null left

Specifications

CAS 167218-46-4
Melting Point 241°C
Color White-Yellow
Molecular Formula C72H54N4
Quantity 1 g
MDL Number MFCD03093703
Synonym TPTE
Formula Weight 975.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diphenyl-N,N′-bis[4′-(diphenylamino)biphenyl-4-yl]benzidine
Show More Show Less