missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diphenyl-4-methoxybenzamide 97.0+%, TCI America™

Catalog No. D17205G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D17205G Supplier TCI America Supplier No. D17205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16034-40-5
Molecular Formula C20H17NO2
Molecular Weight (g/mol) 303.36
MDL Number MFCD00059300
InChI Key GVALSXYCLDABKT-UHFFFAOYSA-N
Synonym N-p-Anisoyldiphenylamine
PubChem CID 584785
IUPAC Name 4-methoxy-N,N-diphenylbenzamide
SMILES COC1=CC=C(C=C1)C(=O)N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 140°C
Color White
Quantity 5 g
Formula Weight 303.36
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diphenyl-4-methoxybenzamide