missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diphenyl-2-naphthylamine 98.0+%, TCI America™

Catalog No. D48431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48431G 1 g
D48435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48431G Supplier TCI America Supplier No. D48431G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6940-30-3
Molecular Formula C22H17N
Molecular Weight (g/mol) 295.385
InChI Key VWYDYNGPYMOCMD-UHFFFAOYSA-N
PubChem CID 235845
IUPAC Name N,N-diphenylnaphthalen-2-amine
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC4=CC=CC=C4C=C3

Specifications

Melting Point 125°C
Color White
Quantity 1 g
Formula Weight 295.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diphenyl-2-naphthylamine