missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenyl-1,4-phenylenediamine 95.0+%, TCI America™

$87.60 - $822.05

Chemical Identifiers

CAS 74-31-7
Molecular Formula C18H16N2
Molecular Weight (g/mol) 260.34
MDL Number MFCD00003015
InChI Key UTGQNNCQYDRXCH-UHFFFAOYSA-N
Synonym n,n'-diphenyl-p-phenylenediamine, n1,n4-diphenylbenzene-1,4-diamine, n,n'-diphenyl-1,4-phenylenediamine, dppd, flexamine g, agerite dppd, diphenyl-p-phenylenediamine, dffd, nonflex h, stabilizer dppd
PubChem CID 6319
ChEBI CHEBI:34860
IUPAC Name N1,N4-diphenylbenzene-1,4-diamine
SMILES N(C1=CC=CC=C1)C1=CC=C(NC2=CC=CC=C2)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D060925G
View Documents
TCI America
D060925G
25 g
Each for $87.60
Only null left
 
D0609100G
View Documents
TCI America
D0609100G
100 g
Each for $240.46
Only null left
 
D0609500G
View Documents
TCI America
D0609500G
500 g
Each for $822.05
Only null left
 
Description

Description

Specifications

Chemical Identifiers

74-31-7
260.34
UTGQNNCQYDRXCH-UHFFFAOYSA-N
6319
N1,N4-diphenylbenzene-1,4-diamine
C18H16N2
MFCD00003015
n,n'-diphenyl-p-phenylenediamine, n1,n4-diphenylbenzene-1,4-diamine, n,n'-diphenyl-1,4-phenylenediamine, dppd, flexamine g, agerite dppd, diphenyl-p-phenylenediamine, dffd, nonflex h, stabilizer dppd
CHEBI:34860
N(C1=CC=CC=C1)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

74-31-7
225°C
MFCD00003015
n,n'-diphenyl-p-phenylenediamine, n1,n4-diphenylbenzene-1,4-diamine, n,n'-diphenyl-1,4-phenylenediamine, dppd, flexamine g, agerite dppd, diphenyl-p-phenylenediamine, dffd, nonflex h, stabilizer dppd
N(C1=CC=CC=C1)C1=CC=C(NC2=CC=CC=C2)C=C1
260.34
CHEBI:34860
≥95.0% (T)
N,N′-Diphenyl-1,4-phenylenediamine
152°C
C18H16N2
25 g
UTGQNNCQYDRXCH-UHFFFAOYSA-N
N1,N4-diphenylbenzene-1,4-diamine
6319
260.34
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0145

RTECSNumber : ST2275000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'