missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenyl-1,4-phenylenediamine 95.0+%, TCI America™

Catalog No. D0609500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0609500G 500 g
D060925G 25 g
D0609100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0609500G Supplier TCI America Supplier No. D0609500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74-31-7
Molecular Formula C18H16N2
Molecular Weight (g/mol) 260.34
MDL Number MFCD00003015
InChI Key UTGQNNCQYDRXCH-UHFFFAOYSA-N
Synonym n,n'-diphenyl-p-phenylenediamine, n1,n4-diphenylbenzene-1,4-diamine, n,n'-diphenyl-1,4-phenylenediamine, dppd, flexamine g, agerite dppd, diphenyl-p-phenylenediamine, dffd, nonflex h, stabilizer dppd
PubChem CID 6319
ChEBI CHEBI:34860
IUPAC Name N1,N4-diphenylbenzene-1,4-diamine
SMILES N(C1=CC=CC=C1)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 152°C
Boiling Point 225°C
Quantity 500 g
Formula Weight 260.34
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diphenyl-1,4-phenylenediamine
EINECSNumber (3)-0145
RTECSNumber ST2275000
TSCA Yes