missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylurea, 98%

Catalog No. AAB2132936
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2132936 500 g
AAB2132930 250 g
AAB213290B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2132936 Supplier Thermo Scientific Chemicals Supplier No. B2132936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96-31-1 | C3H8N2O | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-31-1
Molecular Formula C3H8N2O
Molecular Weight (g/mol) 88.11
MDL Number MFCD00008286
InChI Key MGJKQDOBUOMPEZ-UHFFFAOYSA-N
Synonym n,n'-dimethylurea, sym-dimethylurea, urea, n,n'-dimethyl, dimethylurea, symmetric dimethylurea, urea, 1,3-dimethyl, n,n'-dimethylharnstoff, 1,3-dimethyl urea, n,n'-dimethylharnstoff german, unii-wam6dr9i4x
PubChem CID 7293
ChEBI CHEBI:80472
IUPAC Name 1,3-dimethylurea
SMILES CNC(=O)NC

Specifications

Melting Point 100°C to 104°C
Density 1.14
Boiling Point 268°C to 270°C (decomposition)
Flash Point 157°C (314°F)
Quantity 500 g
Beilstein 1740672
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material N,N'-Dimethylurea
EINECSNumber 202-498-7
RTECSNumber YS9868000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only