missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyltetradecylamine 90.0+%, TCI America™

Catalog No. D184425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D184425ML 25 mL
D1844500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D184425ML Supplier TCI America Supplier No. D184425ML

Chemical Identifiers

CAS 112-75-4
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.463
MDL Number MFCD00053736
InChI Key SFBHPFQSSDCYSL-UHFFFAOYSA-N
Synonym N,N-Dimethylmyristylamine
PubChem CID 8211
IUPAC Name N,N-dimethyltetradecan-1-amine
SMILES CCCCCCCCCCCCCCN(C)C

Specifications

Color Yellow
Boiling Point 148°C
Quantity 25 mL
UN Number 2735
Formula Weight 241.46
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dimethyltetradecylamine
EINECSNumber (2)-0176
TSCA Yes