missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylquinacridone 96.0+%, TCI America™

Catalog No. D26875G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26875G 5 g
D26871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26875G Supplier TCI America Supplier No. D26875G
Only null left

Chemical Identifiers

CAS 19205-19-7
Molecular Formula C22H16N2O2
Molecular Weight (g/mol) 340.382
MDL Number MFCD00683078
InChI Key SCZWJXTUYYSKGF-UHFFFAOYSA-N
Synonym 5,12-Dihydro-5,12-dimethylquino[2,3-b]acridine-7,14-dione, DMQA
PubChem CID 2844779
IUPAC Name 5,12-dimethylquinolino[2,3-b]acridine-7,14-dione
SMILES CN1C2=CC=CC=C2C(=O)C3=CC4=C(C=C31)C(=O)C5=CC=CC=C5N4C

Specifications

Quantity 5 g
Formula Weight 340.38
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dimethylquinacridone