missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylpropionamide 98.0+%, TCI America™
$93.21 - $779.17
Chemical Identifiers
| CAS | 758-96-3 |
|---|---|
| Molecular Formula | C5H11NO |
| Molecular Weight (g/mol) | 101.149 |
| MDL Number | MFCD00009301 |
| InChI Key | MBHINSULENHCMF-UHFFFAOYSA-N |
| Synonym | n,n-dimethylpropionamide, propanamide, n,n-dimethyl, dimethylamide of propionic acid, propionamide, n,n-dimethyl, n,n-dimethyl propionamide, acmc-209p0o, propionic acid dimethylamide, 4-04-00-00184 beilstein handbook reference, ksc494e1j |
| PubChem CID | 12965 |
| IUPAC Name | N,N-dimethylpropanamide |
| SMILES | CCC(=O)N(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D079325ML
|
TCI America
D079325ML |
25 mL |
Each for $93.21
|
|
|||||
|
D0793100ML
|
TCI America
D0793100ML |
100 mL |
Each for $272.16
|
|
|||||
|
D0793500ML
|
TCI America
D0793500ML |
500 mL |
Each for $779.17
|
|
|||||
Chemical Identifiers
| 758-96-3 | |
| 101.149 | |
| MBHINSULENHCMF-UHFFFAOYSA-N | |
| 12965 | |
| CCC(=O)N(C)C |
| C5H11NO | |
| MFCD00009301 | |
| n,n-dimethylpropionamide, propanamide, n,n-dimethyl, dimethylamide of propionic acid, propionamide, n,n-dimethyl, n,n-dimethyl propionamide, acmc-209p0o, propionic acid dimethylamide, 4-04-00-00184 beilstein handbook reference, ksc494e1j | |
| N,N-dimethylpropanamide |
Specifications
| 758-96-3 | |
| 176°C | |
| MFCD00009301 | |
| n,n-dimethylpropionamide, propanamide, n,n-dimethyl, dimethylamide of propionic acid, propionamide, n,n-dimethyl, n,n-dimethyl propionamide, acmc-209p0o, propionic acid dimethylamide, 4-04-00-00184 beilstein handbook reference, ksc494e1j | |
| CCC(=O)N(C)C | |
| 101.149 | |
| 101.15 | |
| Liquid |
| Colorless | |
| C5H11NO | |
| 25 mL | |
| MBHINSULENHCMF-UHFFFAOYSA-N | |
| N,N-dimethylpropanamide | |
| 12965 | |
| ≥98.0% (GC) | |
| N,N-Dimethylpropionamide |
Safety and Handling
EINECSNumber : (2)-3461
RTECSNumber : UE3810000
TSCA : Yes