missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylpropionamide 98.0+%, TCI America™

Catalog No. D0793500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0793500ML 500 mL
D079325ML 25 mL
D0793100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0793500ML Supplier TCI America Supplier No. D0793500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 758-96-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00009301
InChI Key MBHINSULENHCMF-UHFFFAOYSA-N
Synonym n,n-dimethylpropionamide, propanamide, n,n-dimethyl, dimethylamide of propionic acid, propionamide, n,n-dimethyl, n,n-dimethyl propionamide, acmc-209p0o, propionic acid dimethylamide, 4-04-00-00184 beilstein handbook reference, ksc494e1j
PubChem CID 12965
IUPAC Name N,N-dimethylpropanamide
SMILES CCC(=O)N(C)C

Specifications

Color Colorless
Boiling Point 176°C
Quantity 500 mL
Formula Weight 101.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylpropionamide
EINECSNumber (2)-3461
RTECSNumber UE3810000
TSCA Yes