missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylpropionamide, 98+%

Catalog No. p-7056097
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6485014 25 g
AAH6485022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6485014 Supplier Thermo Scientific Chemicals Supplier No. H6485014
Only null left

CAS: 758-96-3 | C5H11NO | 101.149 g/mol

N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.

Solubility
Soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 758-96-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00009301
InChI Key MBHINSULENHCMF-UHFFFAOYSA-N
Synonym n,n-dimethylpropionamide, propanamide, n,n-dimethyl, dimethylamide of propionic acid, propionamide, n,n-dimethyl, n,n-dimethyl propionamide, acmc-209p0o, propionic acid dimethylamide, 4-04-00-00184 beilstein handbook reference, ksc494e1j
PubChem CID 12965
IUPAC Name N,N-dimethylpropanamide
SMILES CCC(=O)N(C)C

Specifications

Melting Point -45°C (literature)
Color Colorless
Density 0.920 g/mL
Boiling Point 174°C to 175°C (literature)
Flash Point 62°C (144°F)
Refractive Index 1.44
Linear Formula CH3CH2CON(CH3)2
Quantity 25 g
Beilstein 1740719
Solubility Information Soluble in water.
Formula Weight 101.15
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material N,N-Dimethylpropionamide
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 212-064-9
RTECSNumber UE3810000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only