missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyloctadecylamine, 89%, tech.

CAS: 124-28-7 | C20H44ClN | 334.03 g/mol

Supplier:  Thermo Scientific Chemicals 408430010

 View more versions of this product

Catalog No. AC408430010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N, N-Dimethyloctadecylamine
124-28-7
100.0
Colorless to Yellow
347.0°C
Authentic
Glass bottle
(CH3)2N(CH2)17CH3
1 kg
15, 3522
Solubility in water: slightly soluble
[Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
334.03
297.57
Technical
tech., 89%
84.0
15.0°C to 20.0°C
0.8000g/mL
155°C
84% min. (C18) (GC)
C20H44ClN
MFCD00048496
0.8
n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
PFKRTWCFCOUBHS-UHFFFAOYSA-N
N,N-dimethyloctadecan-1-amine
15365
89%
Liquid After Melting

Chemical Identifiers

124-28-7
334.03
PFKRTWCFCOUBHS-UHFFFAOYSA-N
15365
[Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
C20H44ClN
MFCD00048496
n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
N,N-dimethyloctadecan-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 204-694-8

RTECSNumber : RG4200000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only