missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyloctadecylamine, 89%, tech.

Catalog No. AC408432500
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408432500 250 g Glass bottle
AC408430010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC408432500 Supplier Thermo Scientific Chemicals Supplier No. 408432500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 124-28-7 | C20H44ClN | 334.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 124-28-7
Molecular Formula C20H44ClN
Molecular Weight (g/mol) 334.03
MDL Number MFCD00048496
InChI Key PFKRTWCFCOUBHS-UHFFFAOYSA-N
Synonym n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
PubChem CID 15365
IUPAC Name N,N-dimethyloctadecan-1-amine
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C

Specifications

CAS Min % 84.0
CAS Max % 100.0
Melting Point 15.0°C to 20.0°C
Color Colorless to Yellow
Density 0.8000g/mL
Boiling Point 347.0°C
Flash Point 155°C
Infrared Spectrum Authentic
Assay Percent Range 84% min. (C18) (GC)
Packaging Glass bottle
Linear Formula (CH3)2N(CH2)17CH3
Quantity 250 g
Specific Gravity 0.8
Merck Index 15, 3522
Solubility Information Solubility in water: slightly soluble
Formula Weight 297.57
Percent Purity 89%
Grade Technical
Physical Form Liquid After Melting
Chemical Name or Material N, N-Dimethyloctadecylamine, tech., 89%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 204-694-8
RTECSNumber RG4200000
TSCA TSCA

RUO – Research Use Only