missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylmethyleneammonium Iodide 97.0+%, TCI America™

Catalog No. p-7126847
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16465G 5 g
D164625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16465G Supplier TCI America Supplier No. D16465G
Only null left

Chemical Identifiers

CAS 33797-51-2
Molecular Formula C3H8IN
Molecular Weight (g/mol) 185.008
MDL Number MFCD00011810
InChI Key VVDUZZGYBOWDSQ-UHFFFAOYSA-M
Synonym eschenmoser's salt, n,n-dimethylmethyleneiminium iodide, n-methyl-n-methylenemethanaminium iodide, dimethyl methylidene azanium iodide, n,n-dimethyl methyleneammonium iodide, dimethyl methylene ammonium iodide, methanaminium, n-methyl-n-methylene-, iodide, eschenmoser inverted exclamation mark s salt, eschenmoser salt, dimethyl methylene ammounium iodide
PubChem CID 2724133
IUPAC Name dimethyl(methylidene)azanium;iodide
SMILES C[N+](=C)C.[I-]

Specifications

Quantity 5 g
Formula Weight 185.01
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethylmethyleneammonium Iodide
TSCA No
Recommended Storage Refrigerator