missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylisopropylamine 99.0+%, TCI America™

Catalog No. D1563500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1563500ML 500 mL
D156325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1563500ML Supplier TCI America Supplier No. D1563500ML

Chemical Identifiers

CAS 996-35-0
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.17
MDL Number MFCD00015014
InChI Key VMOWKUTXPNPTEN-UHFFFAOYSA-N
Synonym n,n-dimethylisopropylamine, dimethylisopropylamine, 2-propanamine, n,n-dimethyl, isopropyldimethylamine, ethylamine, n,n,1-trimethyl, unii-wa4te070j7, 2-dimethylaminopropane, ch3 2 i-c3h7 n, dmipa, n-isopropyldimethylamine
PubChem CID 70452
IUPAC Name dimethyl(propan-2-yl)amine
SMILES CC(C)N(C)C

Specifications

Color Colorless
Boiling Point 68°C
Quantity 500 mL
UN Number 2733
Formula Weight 87.17
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylisopropylamine
EINECSNumber (2)-4066
TSCA Yes