missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylisobutyramide 98.0+%, TCI America™
Supplier: TCI America D491825G
Specifications
| N,N-Dimethylisobutyramide | |
| Yellow | |
| C6H13NO | |
| 25 g | |
| GXMIHVHJTLPVKL-UHFFFAOYSA-N | |
| N,N,2-trimethylpropanamide | |
| 243415 | |
| ≥98.0% (GC) |
| 21678-37-5 | |
| 179°C | |
| MFCD00075378 | |
| n,n-dimethylisobutyramide, n,n,2-trimethylpropionamide, n,n-dimethyl-isobutyramide, acmc-20ak9x, n,n-dimethylisobutylamide, isobutyric acid dimethylamide, propanamide, n,n,2-trimethyl, propanamide,n,n,2-trimethyl, n,n-dimethyl2-methylpropionamide | |
| CC(C)C(=O)N(C)C | |
| 115.176 | |
| 115.18 | |
| Liquid |
Chemical Identifiers
| 21678-37-5 | |
| 115.176 | |
| GXMIHVHJTLPVKL-UHFFFAOYSA-N | |
| 243415 | |
| CC(C)C(=O)N(C)C |
| C6H13NO | |
| MFCD00075378 | |
| n,n-dimethylisobutyramide, n,n,2-trimethylpropionamide, n,n-dimethyl-isobutyramide, acmc-20ak9x, n,n-dimethylisobutylamide, isobutyric acid dimethylamide, propanamide, n,n,2-trimethyl, propanamide,n,n,2-trimethyl, n,n-dimethyl2-methylpropionamide | |
| N,N,2-trimethylpropanamide |
Safety and Handling
TSCA : No