missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylisobutyramide 98.0+%, TCI America™

Catalog No. D49185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49185G 5 g
D491825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49185G Supplier TCI America Supplier No. D49185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21678-37-5
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00075378
InChI Key GXMIHVHJTLPVKL-UHFFFAOYSA-N
Synonym n,n-dimethylisobutyramide, n,n,2-trimethylpropionamide, n,n-dimethyl-isobutyramide, acmc-20ak9x, n,n-dimethylisobutylamide, isobutyric acid dimethylamide, propanamide, n,n,2-trimethyl, propanamide,n,n,2-trimethyl, n,n-dimethyl2-methylpropionamide
PubChem CID 243415
IUPAC Name N,N,2-trimethylpropanamide
SMILES CC(C)C(=O)N(C)C

Specifications

Color Yellow
Boiling Point 179°C
Quantity 5 g
Formula Weight 115.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylisobutyramide