missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylisobutyramide, 99%

Catalog No. p-7035706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5343006 5 g
AAH5343014 25 g
AAH5343022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5343006 Supplier Thermo Scientific Chemicals Supplier No. H5343006
Only null left

CAS: 21678-37-5 | C6H13NO | 115.176 g/mol

1- chloro-N,N-2-trimethylpropenylamine readily prepared' from N,N-dimethylisobutyramide. Isoxazoles are prepared by deprotonation of N,N-dimethylisobutyramide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1- chloro-N,N-2-trimethylpropenylamine readily prepared′ from N,N-dimethylisobutyramide. Isoxazoles are prepared by deprotonation of N,N-dimethylisobutyramide.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 21678-37-5
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00075378
InChI Key GXMIHVHJTLPVKL-UHFFFAOYSA-N
Synonym n,n-dimethylisobutyramide, n,n,2-trimethylpropionamide, n,n-dimethyl-isobutyramide, acmc-20ak9x, n,n-dimethylisobutylamide, isobutyric acid dimethylamide, propanamide, n,n,2-trimethyl, propanamide,n,n,2-trimethyl, n,n-dimethyl2-methylpropionamide
PubChem CID 243415
IUPAC Name N,N,2-trimethylpropanamide
SMILES CC(C)C(=O)N(C)C

Specifications

Density 0.892
Boiling Point 175°C to 176°C (44 mmHg)
Flash Point 154°C (309°F)
Refractive Index 1.439
Linear Formula (CH3)2CHCON(CH3)2
Quantity 5 g
Beilstein 1742178
Solubility Information Soluble in water.
Formula Weight 115.18
Percent Purity 99%
Chemical Name or Material N,N-Dimethylisobutyramide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only