missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylhexadecylamine 98.0+%, TCI America™

Catalog No. D1568500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1568500ML 500 mL
D156825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1568500ML Supplier TCI America Supplier No. D1568500ML

Chemical Identifiers

CAS 112-69-6
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.517
MDL Number MFCD00015086
InChI Key NHLUVTZJQOJKCC-UHFFFAOYSA-N
Synonym 1-(Dimethylamino)hexadecane, N,N-Dimethylcetylamine, N,N-Dimethylpalmitylamine
PubChem CID 16221
IUPAC Name N,N-dimethylhexadecan-1-amine
SMILES CCCCCCCCCCCCCCCCN(C)C

Specifications

Color Colorless
Boiling Point 148°C
Quantity 500 mL
UN Number 2735
Formula Weight 269.52
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylhexadecylamine
EINECSNumber (2)-0176&(2)-0185
RTECSNumber MM0750000
TSCA Yes