missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylglycine Methyl Ester 98.0+%, TCI America™

Catalog No. D0257500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0257500G 500 g
D025725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0257500G Supplier TCI America Supplier No. D0257500G

Chemical Identifiers

CAS 7148-06-3
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00059303
InChI Key LRZFEBJUJIQVDQ-UHFFFAOYSA-N
Synonym n,n-dimethylglycine methyl ester, methyl 2-dimethylamino acetate, glycine, n,n-dimethyl-, methyl ester, methyl dimethylamino acetate, methyl n,n-dimethylglycinate, acmc-2097kq, dimethylglycine methyl ester, ch3 2nch2cooch3, dimethylaminoessigsauremethylester
PubChem CID 81569
IUPAC Name methyl 2-(dimethylamino)acetate
SMILES CN(C)CC(=O)OC

Specifications

Color Colorless
Boiling Point 135°C
Quantity 500 g
UN Number 3272
Formula Weight 117.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylglycine Methyl Ester
RTECSNumber MC0011000
TSCA No
Recommended Storage Freezer