missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylglycine methyl ester, 98%

Catalog No. AAL1189706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1189706 5 g
AAL1189714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1189706 Supplier Thermo Scientific Chemicals Supplier No. L1189706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7148-06-3 | C8H11ClNO2 | 188.63 g/mol

N,N-Dimethylglycine methyl ester is used as a biochemical for proteomics research. It is also used as chemical and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylglycine methyl ester is used as a biochemical for proteomics research. It is also used as chemical and pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
It is sensitive to air and hygroscopic in nature. Protect from humidity and water. Store under dry inert gas. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 7148-06-3
MDL Number MFCD00059303

Specifications

Boiling Point 134°C to 135°C
Molecular Formula C5H11NO2
Refractive Index 1.412
Linear Formula (CH3)2NCH2CO2CH3
UN Number UN3272
Beilstein 1744788
Synonym n,n-dimethylglycine methyl ester, methyl 2-dimethylamino acetate, glycine, n,n-dimethyl-, methyl ester, methyl dimethylamino acetate, methyl n,n-dimethylglycinate, acmc-2097kq, dimethylglycine methyl ester, ch3 2nch2cooch3, dimethylaminoessigsauremethylester
Solubility Information Soluble in water.
InChI Key LRZFEBJUJIQVDQ-UHFFFAOYSA-N
Density 0.95
SMILES CN(C)CC(=O)OC
IUPAC Name methyl 2-(dimethylamino)acetate
Quantity 5 g
Molecular Weight (g/mol) 117.15
PubChem CID 81569
Flash Point 38°C (100°F)
Formula Weight 117.15
Sensitivity Air sensitive; Hygroscopic
Percent Purity 98%
Chemical Name or Material N,N-Dimethylglycine methyl ester
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
RTECSNumber MC0011000
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only