missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylglycine Hydrochloride, Spectrum™ Chemical

Catalog No. 18611884
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611884 25 g Poly Bottle
18611881 100 g Poly Bottle
18611885 500 g Poly Bottle
18611882 2.5 kg Poly Bottle
18611883 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611884 Supplier Spectrum Chemical Mfg Cor Supplier No. D118025GM

Chemical Identifiers

CAS 2491-06-7

Specifications

CAS Max % 1
Melting Point 188° to 194°C
Packaging Poly Bottle
Quantity 25 g
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.