missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, Spectrophotometric Grade, 99.7+%

Catalog No. p-4408417
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA13808K2 1 L
AA13808M1 4 x 1 L
AA13808K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA13808K2 Supplier Thermo Scientific Chemicals Supplier No. 013808K2
Only null left

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1% | CAS: 68-12-2 | C3H7NO | 73.10 g/mol

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films. It is suitable for use in ultraviolet spectrophotometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Color Colorless
Density 0.944g/cm3
Ignition Point 440°C (824°F)
Odor Amine-like
Assay Percent Range >99.7%
Quantity 1 L
Vapor Pressure 3.5 hPa (3mm Hg) at 20°C
Formula Weight 73.09
Grade Sprectrophotometric
Physical Form Liquid
Chemical Name or Material N, N-Dimethylformamide, Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%
Show More Show Less
Hazard Category H360-H226-H312-H332-H319: May damage fertility or the unborn child. Flammable liquid and vapour. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation.
DOTInformation Hazard Class: 3, Packing Group: III
EINECSNumber 200-679-5

RUO – Research Use Only