missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.8% (GC),Solstice

Catalog No. 60046196
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
180 kg
Packaging:
Glass Bottle
Metal Drum
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046196 1 L Glass Bottle
60046197 2.5 L Glass Bottle
60046198 2.5 L Poly Bottle
60046201 6 x 1 L Glass Bottle
60046200 4 x 2.5 L Glass Bottle
60046195 180 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60046196 Supplier Solstice Supplier No. 331201L

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.8% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

pH 7 (at 20°C)
Melting Point -61°C
Density 0.944g/mL, 0.948 to 0.950g/mL at 20°C
Quantity 1 L
Explosion Limit 0.152
Flash Point 58°C (136.4°F)
Ignition Point 833°F
Odor Weak
Packaging Glass Bottle
Physical Form Liquid
Linear Formula HCON(CH3)2
Vapor Pressure 2.7mmHg (20°C)
UN Number UN2265
Beilstein 605365
Boiling Point 152°C to 154°C
Formula Weight 73.09g/mol
Percent Purity ≥99.8% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material N,N-Dimethylformamide, Reag. Ph. Eur.
Show More Show Less
Hazard Category Danger
Hazard Statement P201-P210-P261-P280-P308 + P313-P370 + P378
Precautionary Statement H226-H312 + H332-H319-H360D
DOTInformation 3/PGIII
EINECSNumber 200-679-5
RTECSNumber LQ2100000