missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.8% (GC),Honeywell™ Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.8% (GC)
Supplier: Honeywell-Riedel-de Haen 33120180KG
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
| 7 (at 20°C) | |
| -61°C | |
| 68-12-2 | |
| 152°C to 154°C | |
| 0.152 | |
| 833°F | |
| Metal Drum | |
| C3H7NO | |
| MFCD00003284 | |
| UN2265 | |
| dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
| CN(C)C=O | |
| 73.10 | |
| CHEBI:17741 | |
| ≥99.8% (GC) |
| N,N-Dimethylformamide | |
| Reag. Ph. Eur. | |
| 0.944g/mL, 0.948 to 0.950g/mL at 20°C | |
| 180 kg | |
| 58°C (136.4°F) | |
| Weak | |
| Liquid | |
| HCON(CH3)2 | |
| 2.7mmHg (20°C) | |
| 605365 | |
| ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
| N,N-dimethylformamide | |
| 6228 | |
| 73.09g/mol | |
| ACS/puriss. p.a. |
Chemical Identifiers
| 68-12-2 | |
| 73.10 | |
| ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
| 6228 | |
| N,N-dimethylformamide |
| C3H7NO | |
| MFCD00003284 | |
| dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
| CHEBI:17741 | |
| CN(C)C=O |
Safety and Handling
P201-P210-P261-P280-P308 + P313-P370 + P378
H226-H312 + H332-H319-H360D
Danger
DOTInformation : 3/PGIII
EINECSNumber : 200-679-5
RTECSNumber : LQ2100000