missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide, Purified (DMF), Reagents
Specifications
Specifications
| Melting Point | -60.5°C |
| Color | Colorless to Slight Yellow |
| Quantity | 1 L |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.944 |
| Boiling Point | 152°C to 153°C |
| Grade | Pure |
| Chemical Name or Material | N,N-Dimethylformamide, Purified (DMF) |